Learn More
Alpco Ltd Cortisol ELISA (Saliva)
SDP
Supplier:  Alpco Ltd 11CORHUE01SLV
Encompass
Cortisol ELISA (Saliva) is an enzyme immunoassay for the quantitative determination of cortisol in human saliva.
  • Includes ready to use standards and controls
  • No cross reaction with DHEAS and tetrahydrocortisone
  • Incubation time of 60-65 minutes
For research use only. Not for use in diagnostic procedures.
Catalog No. 50-921-686