Learn More
Alkali Scientific ASI™ Direct Thermal Cryo Label, 38x19mm for Lg Vials/Tubes, 750 per Roll, 1/Roll, White
SDP
Supplier:  Alkali Scientific LDTC11234W
Encompass_Preferred
DIRECT THERMAL CRYO LABEL 38x19mm for Lg Vials/Tubes We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Labels Per case: 750, 1 Roll/cs
Catalog No. 50-302-7621