Learn More
GA International GA International Thermal-Transfer / Direct Thermal Desktop Printer #Cody-3
Supplier:  GA International CODY3
Encompass
Cody-3™ GA International Thermal-Transfer / Direct Thermal Desktop PrinterPrint Method: Thermal Transfer/Direct ThermalResolution: 300 dpiPrint Speed up to: 4″/s (102 mm/s)Print Width up to: 4.16″ (105.7 mm)Label Width: 0.79″ – 4.64″ (20 – 118 mm)Label Height: 0.16″– 30.0″ (4 – 762mm)Label Material Thickness: 0.003″ – 0.01″ (0.076 – 0.2 mm)Liner Material Width: 0.79″ – 4.64″ (20 – 118 mm)Roll Outer Diameter: 5″ (127 mm)Roll Core Diameter: 1″ – 1.5″ (25.4 – 38.1 mm)Interfaces: USB 2.0, Serial (RS232), Ethernet 10/100, USB Host
Catalog No. NC2740876