Learn More
Zymo Research Corporation Direct-zol™ RNA MiniPrep (200 Preps) w/ Zymo-Spin™ IIC Columns (Capped) (Product Supplied w/ 200 ml TRI Reagent™) [Includes R2052 x 1, R2050-1-200 x 1 -packaged separately]
SDP
Supplier:  Zymo Research Corporation R2053
Encompass_Preferred
Total RNA including small RNAs (17-200 nt)2, is effectively isolated from a variety of sample sources (cells, tissues, serum, plasma, blood, samples stored in DNA/RNA Shield™, etc.).Simply add ethanol to a TRI Reagent sample, bind directly to the Zymo-Spin™ Column, wash, and elute RNA. No phase separation, precipitation, or post-purification steps are necessary. RNA is high-quality and ready for Next-Gen Sequencing, RTqPCR, transcription profiling, hybridization, etc.
Catalog No. 50-444-629