Learn More
Zymo Research Corporation Direct-zol™ RNA PreWash (Concentrate) (160 ml)
SDP
Supplier:  Zymo Research Corporation R20502160
Encompass_Preferred
Direct-zol™ RNA Pre-wash (160 ml) for use with Direct-zol™ RNA Purification Kits for rapid isolation of high-quality total RNA including small RNAs (>17 nt) from samples in TRI-Reagent, TRIzol or similar.
Catalog No. 50-444-625