Learn More
Medchemexpress LLC Dirithromycin (LY237216) | 62013-04-1 | 98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0643100MG
Encompass_Preferred
Dirithromycin (LY237216) is a derivative of Erythromycin and functions as a potent and orally active semi-synthetic macrolide antibiotic. It demonstrates activity against gram-positive bacteria, as well as specific species such as *Legionella spp.*, *Helicobacter pylori*, and *Chlamydia trachomatis*.
  • Potent and orally active semi-synthetic macrolide antibiotic.
  • Active against gram-positive bacteria.
  • Active against *Legionella spp.*.
  • Active against *Helicobacter pylori*.
  • Active against *Chlamydia trachomatis*.
  • Exhibits an in vitro spectrum of antimicrobial activity similar to Erythromycin.
  • Effective against experimental infections caused by *S. aureus*, *S. pyogenes*, and *S. pneumoniae* in mice.
Catalog No. 50-004-09249