Learn More
TARGETMOL CHEMICALS INC DIRLOTAPIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T150065MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Dirlotapide (CP742033) is an intestinal selective microsomal triglyceride transfer protein (MTP) inhibitor. It reduces body weight in diabetic dogs.Dirlotapide reduces food intake in a dose-dependent manner possibly by increasing the release of peptide YY into the circulation. purity: 99%
Catalog No. 50-258-1261