Learn More
Medchemexpress LLC HY-100375 100mg Medchemexpress, Diroximel fumarate CAS:1577222-14-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100375100MG
Encompass_Preferred
Medchemexpress, HY-100375 100mg Diroximel fumarate CAS:1577222-14-0 Diroximel fumarate (DRF) is a prodrug of monomethyl fumarate in a controlled-release formulation that rapidly and efficiently converts to MMF in the body [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1798