Learn More
Carbosynth LLC. Fmoc-Asn[per-O-Boc-(Fucα1-6GlcNAcß)], 10mg, MF= C58H81N3O24
SDP
Supplier:  Carbosynth LLC. CCK2210510MG
Encompass_Preferred
Nα-9-Fluorenylmethoxycarbonyl-Nomega-[2,3,4-tri-O-t-Butyloxycarbonyl-α-L-Fucopyranosyl-(1→6)-3,4-di-O-t-Butyloxycarbonyl-2-Acetamido-2-Deoxy-β-D-Glucopyranosyl]-L-Asparagine, Application: Boc-Protected Disaccharide Derivative for N-Linked Glycopeptide Synthesis;Fmoc-Amino Acid for the Synthesis of Fucα(1-6)GlcNAc-β-Asn-Containing Peptide, Notes: For the chemical synthesis of glycopeptides, acetyl group has been widely used for the protection of hydroxy groups in sugar moieties. Scientists at The Noguchi Institute recently developed Boc-protected disaccharide building block which does not require troublesome acetyl deprotection in basic condition. Using this derivative, N-linked glycopeptide synthesis can be achieved moreconveniently and efficiently by standard Fmoc-SPPS., MW: 1204.3
Catalog No. 50-195-2400