Learn More
Apexbio Technology LLC DiSC3(5) 53213-94-8 25mg
SDP
Supplier:  Apexbio Technology LLC A467225
Encompass_Preferred
DiSC3 5 (CAS 53213-94-8) is a fluorescent probe targeting mitochondrial membrane potential and mitochondrial NAD-dependent respiration It is designed to monitor transmembrane potential changes thereby assessing mitochondrial function DiSC3 5 exerts its biological activity primarily through specific interference with mitochondrial NAD-related respiratory pathways In cell-based studies DiSC3 5 demonstrates inhibitory activity with an IC50 value of 8 M in mitochondrial NAD-dependent respiratory systems In the presence of Na /K -ATPase inhibitor ouabain DiSC3 5 induces plasma membrane hyperpolarization in Ehrlich ascites tumor cells Based on these pharmacological properties DiSC3 5 holds research potential in mitochondrial membrane potential detection and cellular bioenergetics studies
Catalog No. 50-000-93505