Learn More
Medchemexpress LLC DiSC3(5) | 53213-94-8 | 99.4% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYD008550MG
Encompass_Preferred
DiSC3(5) is a fluorescent probe utilized as a tracer dye to assess mitochondrial membrane potential. It features an excitation/emission wavelength up to 622/670 nm and can inhibit the respiratory system linked with mitochondrial NAD. In specific conditions, it can induce membrane hyperpolarization.
  • Evaluates mitochondrial membrane potential.
  • Functions as a fluorescent tracer dye.
  • Provides excitation/emission wavelength up to 622/670 nm.
  • Inhibits the respiratory system associated with mitochondrial NAD.
  • Induces membrane hyperpolarization of Ehrlich ascites tumor cells in presence of specific inhibitors.
Catalog No. 50-004-10735