Learn More
Diapharma Group, Inc. Volition Nu Q® Discover H3.1 ELISA Research Use Only Kit
SDP
Supplier:  Diapharma Group, Inc. 10010103
Encompass
Volition’s Nu.Q® Discover H3.1 ELISA Research Use Only kit detects quantitative levels of circulating intact nucleosomes containing Histone H3 in plasma. Nucleosomes are repeating subunits of DNA and histone proteins that constitute human chromatin. During cellular damage, such as apoptosis or necrosis, chromatin is fragmented into oligo- or mono- nucleosomes which can be released into the blood stream.
Catalog No. NC2892761