Learn More
Medchemexpress LLC Disodium hydrogen phosphate | 7558-79-4 | 99.2% | 141.96 g·mol⁻¹ | Na2HPO4 | 50g
SDP
Supplier:  Medchemexpress LLC HYY0308E50G
Encompass_Preferred
Sodium phosphate dibasic (Disodium hydrogen phosphate) for cell culture is a buffer and chelating agent Sodium phosphate dibasic is used to adjust the pH of liquids Sodium phosphate dibasic can be used in cell culture[1]
Catalog No. 50-002-69804