Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Disodium monofluorophosphate | 10163-15-2 | 98.3% | 5 G
SDP

Supplier:  Medchemexpress LLC HYB05405G

Encompass_Preferred

Disodium monofluorophosphate is a biochemical reagent suitable for life science research applications, including use as a biological material or organic compound. It is also utilized as a laboratory chemical and in the manufacturing of various substances.

  • Appearance: white to off-white solid
  • Molecular weight: 143.95
  • Formula: FNa2O3P
  • CAS number: 10163-15-2
  • Stable under recommended storage conditions
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Purity of 98.3%

Catalog No. 50-004-06887


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.