Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Disodium terephthalate | 10028-70-3 | MFCD00013137 | 99.9% | 210.09 g/mol | C8H4Na2O4 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW25012710G

Encompass_Preferred

Disodium terephthalate is the disodium salt of terephthalic acid supplied as a high-purity research chemical for laboratory use. It serves as a building block and precursor in polyester (PET) synthesis and is used as a reagent in biochemical assays and model systems. The material is provided as a fine solid with defined analytical properties suitable for both analytical and preparative applications.

  • High purity: 99.93%
  • Molecular formula C8H4Na2O4; molecular weight 210.09 g/mol
  • Solid form suitable for analytical and preparative work
  • Applications include polyester precursor and biochemical assay reagent
  • Available in small pack sizes for laboratory research, e.g. 10 G

Catalog No. 50-002-82338


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.