Learn More
Research Products International Corp Glass Pasteur Pipets
SDP
Supplier:  Research Products International Corp 248610A
Encompass
High quality flint glass Pasteur pipets are ideal for inoculating culture media and transferring samples and reagents. Pipets can be used with aspirating bulbs or attached to a vacuum apparatus.
Catalog No. 50-238-8405