Learn More
Cole-Parmer Microcaps 1-000-0150 Disposable Glass Micropipettes, 15 uL; 100/Pk
Supplier:  Cole-Parmer 0795025
Encompass_Preferred
Precision-bore glass-capillary tube is topped with a dispensing bulb. Capillary action moves the fluid up the tube. Squeeze the bulb to release fluid. Accurate to ±1% of rated volume. Each pack contains one bulb unit and 100 disposable tubes. Shipping weight 1 lb (0.5 kg).
Catalog No. NC0510618