Learn More
Yamazen Science Inc Disposable Column, Packed with High Performance Silica Gel, Luer Lock Type, 10/Pack
Supplier:  Yamazen Science Inc UW00901
Encompass
This item has a minimum qty of 3 per supplier requirements.
Disposable Polypropylene Column with High Throughput Purification Performance, Packed with High Performance Silica GelPore Size 60 angstroms, Particle Size 40-63 microns, Luer Lock TypeSize: S10/pk
Catalog No. NC1465371