Learn More
Tecan 200uL Disposable Tips - Filtered, Pure, Hanging
Supplier:  Tecan 30000629
Encompass_Preferred
Genuine Tecan 200 ul disposable automation tips for Freedom EVO Liquid Handling Arm™ (LiHa) or Fluent Flexible Channel Arm™ (FCA). Hanging tip format, filtered, conductive. Tecan PURE purity level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors. 24 x 96-tip hanging racks, 2,304 tips/sales unit. LiHa Disposable Tips have been carefully designed and validated to work specifically with Liquid Handling Arms™ on all Tecan platforms. They are produced to stringent standards of quality and reproducibility, ensuring reliable liquid handling performance. Manufacturing is fully automated in a controlled environment, using state-of-the-art production processes, integrated quality control procedures and premium raw materials. During production, each tip undergoes in-line optical inspection using a CCD camera and image recognition software, checking all functional parts to ensure consistent pipetting accuracy and precision.
Catalog No. NC9800244