Learn More
Wilmad Labglass DISPENSER TILT 10ML 500ML COMP
Supplier:  Wilmad Labglass LG7915110
Encompass_Preferred
Dispenser, Tilting, Complete. Capacity: 10mL; Flask Capacity: 500mL; Joint Size: [ST] 24/40. Fixed volume dispenser designed to deliver measured volumes quickly w/ reproducibly.
Catalog No. 50-804-143