Learn More
Wilmad Labglass DISPENSER TILT 25ML/1L COMP
Supplier:  Wilmad Labglass LG7915118
Encompass_Preferred
Dispenser, Tilting, Complete. Capacity: 25mL; Flask Capacity: 1000mL; Joint Size: [ST] 24/40. Fixed volume dispenser designed to deliver measured volumes quickly w/ reproducibly.
Catalog No. 50-805-329