Learn More
Wilmad Labglass DISPENSER TILT 5ML/250ML COMP
Supplier:  Wilmad Labglass LG7915108
Encompass_Preferred
Dispenser, Tilting, Complete. Capacity: 5mL; Flask Capacity: 250mL; Joint Size: [ST] 24/40. Fixed volume dispenser designed to deliver measured volumes quickly w/ reproducibly.
Catalog No. 50-804-632