Learn More
Essendant DISPENSER3.5GWTTSK2/CS
Supplier:  Essendant KCC09361
Encompass_Preferred
WypAll WetTask Customizable Wet Wiping System Bucket is designed for use with the WypAll Power Clean WetTask Wipers for Solvents 06006 (sold separately) This enclosed bucket allows you to add the solvent solution you prefer It s simple hygienic and safe The enclosed system also helps eliminate exposure to chemical vapors and splashes The WetTask closed bucket and wet wiping system can cut solvent usage by 20pct and reduce VOC emissions for a safer more cost-effective workplace Using this system also eliminates the need for laundering dirty contaminated towels and cloth rags Choose WypAll WetTask Customizable Wet Wiping System and take your wiping efficiency to the next level Capacity Range (Volume) [Max]- 3 5 gal Material(s)- Plastic Color(s)- Blue Wringer- Enclosed
Catalog No. 50-000-31582