Learn More
Medchemexpress LLC Disperse Orange 1 | 2581-69-3 | 96.6% | 50 MG

Supplier: Medchemexpress LLC HYD033250MG
Disperse Orange 1 is an orange-orange dye commonly used for fiber dyeing. This chemical compound is also utilized as a fluorescent dye. It appears as a solid with a color ranging from purple to purplish red.
- Functions as a fluorescent dye
- Suitable for fiber dyeing applications
- Appears as a solid
- Color: purple to purplish red
- Molecular weight: 318.33
- Chemical formula: C18H14N4O2
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.