Learn More
Thomas Scientific 96 Pin Sterile Replicator, Long Pin 25mm, 72/Cs, 25mm Long, 1mm Diameter, Packaged 6/Bag
SDP
Supplier:  Thomas Scientific 1149C51
Encompass
The 96 Long Pin Replicator is a disposable pin replicator for one time use, that virtually eliminates the threat of cross-contamination from repeat use metal replicators.
Catalog No. NC9584102
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.