Learn More
Med Vet International Disposable Anesthesia Mask, Vertical ID, J0298DB, 2in x 2.5in, Each
Supplier:  Med Vet International JOR298DB
Encompass_Preferred
A crystal clear plastic material with inflation/deflation valve for better patient fitting. Inexpensive. 15mm OD fitting. Five different sizes. This one-way valve inflates with any standard syringe.
Catalog No. 50-005-83004