Learn More
Lakeland Industries Disposable Aprons
Supplier:  Lakeland Industries CTL601
Encompass_Preferred
MicroMax NS uses a high quality microporous PE film laminate to combine effective splash and particle resistance combined with softness, flexibility and high moisture vapor transmission rate (MVTR) for additional comfort. Fabric chieves the highest classes in all four tests required by the EN 14126 standard so is also suitable for viral bacterial protection such as contaminated blood and body fluids. Apron with neck loop and waist ties. Offers Partial Body Type 6[PB] protection.
Catalog No. 50-284-1073