Learn More
World Precision Instrument Disposable Dissection Boards-WP-630-2
SDP
Supplier:  World Precision Instrument WP6302
Encompass
The DispoCut™ Disposable Dissection Board is strong but inexpensive enough to throw away. It is a clean, safer and more efficient way of handling infectious tissue specimens. Ideal for pathology labs, medical schools, classroom dissections, forensic labs, medical research labs, pharmaceutical labs, surgery and more. When sterilizing, DispoCut™ may be safely wiped down with alcohol or autoclaved. Reversible.
Catalog No. 50-253-8392