Learn More
Lakeland Industries Disposable FR Coverall
Supplier:  Lakeland Industries 7428B2X
Encompass_Preferred
Lakeland Pyrolon Plus 2 coveralls are breathable garments that repel water, grease, oil, and other hazards where flash fire is a concern. 7428B is a secondary FR coverall configured with a hood, elastic wrists, and ankles. Coming in a case of 25, with sizes SM-5X are available, Pyrolon Plus 2 is designed to protect your primary FR from hazardous and non-hazardous substances.
Catalog No. 50-284-0902