Learn More
RENICK LABS LLC Disposable, Parylene-encapsulated stir bars for 8x30mm glass shell vial work, 1000/pk
Supplier:  RENICK LABS LLC RL 352A
Encompass
Need disposable stir bars for use with 8mm glass shell vials? RL 352A stir bars are the perfect size. Parylene is vapor deposited to the surface of every stir bar ensuring the best possible encapsulation and guards against reaction interference. While the vast majority of chemical reactions do not affect the Parylene coating, extended stirring times of heterogeneous mixtures at elevated temperatures, ethereal solvents such as Dioxane and THF may cause degradation.
Catalog No. NC3877348