Learn More
Sigma Aldrich Fine Chemicals Biosciences Disposable UV-grade methac
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z1879921PAK
Encompass
Economical UV-grade methacrylate disposable cuvettes. Select either grooved non-optical walls for better grip or four-sided clear walls. All cells have a 10 mm pathlength and are supplied without stoppers.
Catalog No. 50-290-1860