Learn More
Wilmad Labglass DIST COL HEMPEL 19X500 24/40
Supplier:  Wilmad Labglass LG5792124
Encompass
Distilling Column, Hemple, Jacketed, Silvered. Column ID: 19mm; Length For Packing Material: 500mm; Joint Size: 24/40. Similar to LG-5791-series, but w/ jacket completely silvered except For two narrow clear strips 180degree apart.
Catalog No. 50-800-645