Learn More
Wilmad Labglass DIST COL SNYDER 6 BULB 24/40
Supplier:  Wilmad Labglass LG5850102
Encompass_Preferred
Distilling Column, Snyder. Number of Sections: 6; Overall Length: 475mm; Joint Size: 24/40. Designed For rapid Solvent evaporation while retaining a high percentage of solute. Used as a component in Kuderna-Danish evaporative concentrators.
Catalog No. 50-804-003
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.