Learn More
Chemglass Life Sciences Distilling Adapter, Double, 10° Angled Side Neck, 24/40 Joint, 6mm PTFE Stopcock, 1/2" Swagelok
SDP
Supplier:  Chemglass Life Sciences CG123580
Encompass_Preferred
Distilling Adapter, Double, 10¿ Angled Side Neck, 24/40 Joint, 6mm PTFE Stopcock, 1/2" Swagelok. Adapter is designed for use with flasks or reaction vessel lids that have a 10 or 20¿ angled side neck. Adapter has a lower inner joint and two top outer joints. Lower stopcock is either a 6mm or a 10mm PTFE stopcock with a 1/2¿ or 3/4¿ OD distillate takeoff for use with a compression fitting and TFE tubing. PLEASE NOTE: ADAPTER CAN BE USED WITH COMPOUND ANGLED SIDE NECKS.
Catalog No. 50-153-1062