Learn More
Chemglass Life Sciences Chemglass Distilling Column, Vacuum-Jacketed, 45/50 Outer to 65/40 O-Ring Ball Joint
SDP
Supplier:  Chemglass Life Sciences CG1968D302
Encompass_Preferred
This vacuum-jacketed distilling column is designed for use in distillation assemblies and reactor distillation cart systems. It provides a secure transition from a 45/50 outer joint (top) to a 65/40 O-ring ball joint (bottom), and is intended for use with compatible distillation kits and process reactor setups.
  • Vacuum jacketed construction for improved thermal control and reduced heat loss.
  • 45/50 outer joint (top) to 65/40 O-ring ball joint (bottom) for secure, standardized connections.
  • Designed for installation in distillation assemblies and distillation cart systems.
  • Compatible with standard reactor distillation kit configurations and cart-mounted distillation systems.
Catalog No. 50-273-3553