Learn More
Medchemexpress LLC Disuccinimidyl sulfoxide | 1351828-03-9 | MFCD30497419 | 99.8% | 388.35 g·mol⁻¹ | C14H16N2O9S | 100MG
SDP
Supplier:  Medchemexpress LLC HY123740100MG
Encompass_Preferred
Disuccinimidyl sulfoxide (DSSO) is a homobifunctional, amine-targeting sulfoxide cross-linker designed for mass-spectrometry-based protein cross-linking and interaction mapping. Its CID-cleavable bonds produce characteristic fragmentation patterns that simplify identification of cross-linked peptides, making it suitable for studies of peptides, proteins, and multisubunit complexes.
  • MS-cleavable cross-linker that aids peptide identification
  • Homobifunctional, amine-reactive chemistry for lysine targeting
  • CID-cleavable sites generate characteristic fragmentation patterns
  • Suitable for peptides, proteins, and multisubunit complexes
  • Supplied as a crystalline solid for solution preparation
  • High purity for analytical mass spectrometry
Catalog No. 50-002-36589