Learn More
Medchemexpress LLC Disuccinimidyl sulfoxide | 1351828-03-9 | MFCD30497419 | 99.8% | 388.35 g/mol | C14H16N2O9S | 1G
SDP
Supplier:  Medchemexpress LLC HY1237401G
Encompass_Preferred
Disuccinimidyl sulfoxide (DSSO) is a homobifunctional, amine-reactive sulfoxide-containing cross-linker developed for protein and peptide cross-linking in mass spectrometry workflows. Its symmetric CID-cleavable sites generate characteristic fragmentation patterns that simplify identification of cross-linked peptides and support structural proteomics studies.
  • Homobifunctional, amine-reactive NHS ester chemistry.
  • MS-cleavable with symmetric CID-cleavable sites for simplified identification.
  • Suitable for mapping protein-protein interactions and multisubunit complexes.
  • Provided as a solid, commonly supplied in a 1 g size.
  • High purity suitable for proteomics applications.
  • Molecular weight 388.35 g/mol; chemical formula C14H16N2O9S.
Catalog No. 50-002-36590