Learn More
Cayman Chemical DIsuccInImIdyl SulfoxIde 250mg
SDP
Supplier:  Cayman Chemical 9002863250 MG
Encompass_Preferred
A protein cross-linking probe designed to be used with MS; contains two symmetric collision-induced dissociation-cleavable sites that allow effective identification of DSSO-cross-linked peptides based on their distinct fragmentation patterns
Catalog No. 50-289-4441