Learn More
Apexbio Technology LLC Disulfiram, 5g. Cas: 97-77-8 MFCD: MFCD00009048. Dopamine β-hydroxylase inhibitor
SDP
Supplier:  Apexbio Technology LLC A40155000
Encompass_Preferred
Disulfiram blocks the processing of alcohol in the body by inhibiting acetaldehyde dehydrogenase thus causing an unpleasant reaction when alcohol is consumed. Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8518