Learn More
Medchemexpress LLC Dithiodipropionic acid | 1119-62-6 | MFCD00002780 | 99.0% | 210.26 | C6H10O4S2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01439510G
Encompass_Preferred
Dithiodipropionic acid is a disulfide-containing dicarboxylic acid used as a building block in polymer synthesis, bioconjugation, and general laboratory research. It is supplied as a white to off-white solid with high purity and defined storage recommendations to preserve stability.
  • Contains a reducible disulfide linkage useful for redox-responsive polymers and linkers.
  • Dicarboxylic functionality enables esterification and amide coupling.
  • High purity suitable for analytical and synthetic applications.
  • Supplied in laboratory-scale packaging for convenient use.
  • Stable as a powder with recommended cold storage to extend shelf life.
Catalog No. 50-003-03805