Learn More
Medchemexpress LLC Dithiodipropionic acid | 1119-62-6 | MFCD00002780 | 99.0% | 210.26 g/mol | C6H10O4S2 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYW014395
Encompass_Preferred
Dithiodipropionic acid is a disulfide-containing dicarboxylic acid used as a research reagent and synthetic building block. It is supplied as a white to off-white solid, has a molecular weight of about 210.26, and is commonly used where a reducible disulfide linkage is required in conjugation, formulation, or materials research.
Catalog No. 50-003-02416