Learn More
Medchemexpress LLC Dithiooxamide | 79-40-3 | 99.9% | 120.20 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0104881G
Encompass_Preferred
Dithiooxamide (Rubeanic Acid) is a selective chelating agent and a sulfur-containing analog of Oxamide. It forms stable chelates with various metal ions (such as lead, iron, cadmium, manganese, and aluminum), which can be separated, enriched, and detected by adsorption on solid adsorbents or electrode surfaces.
  • Selective chelating agent
  • Sulfur-containing analog of Oxamide
  • Forms stable chelates with various metal ions
Catalog No. 50-003-67056