Learn More
Medchemexpress LLC Dithiooxamide | 79-40-3 | 99.9% | 120.20 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01048810G
Encompass_Preferred
Dithiooxamide (Rubeanic Acid) is a selective chelating agent and a sulfur-containing analog of Oxamide. It forms stable chelates with various metal ions, including lead, iron, cadmium, manganese, and aluminum, allowing for their separation, enrichment, and detection by adsorption on solid adsorbents or electrode surfaces.
  • Acts as a selective chelating agent
  • Forms stable chelates with various metal ions
  • Enables separation, enrichment, and detection of metal ions
  • Suitable for use in biochemical assays
Catalog No. 50-004-53791