Learn More
Medchemexpress LLC Dithiooxamide (rubeanic acid) | 79-40-3 | 99.9% | 120.20 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01048850G
Encompass_Preferred
Dithiooxamide (Rubeanic Acid) is a selective chelating agent and a sulfur-containing analog of Oxamide. It forms stable chelates with various metal ions such as lead, iron, cadmium, manganese, and aluminum. These chelates can be separated, enriched, and detected through adsorption on solid adsorbents or electrode surfaces.
  • Acts as a selective chelating agent.
  • Sulfur-containing analog of oxamide.
  • Forms stable chelates with various metal ions.
  • Chelates can be separated, enriched, and detected by adsorption.
Catalog No. 50-004-53911