Learn More
Abcam Dithiothreitol (DTT), reducing reagent, 1ML
Supplier:  Abcam AB1413901ML
Encompass_Preferred
MW 154.3 Da, Purity >99%. Cell-permeable reducing reagent. Prevents oxidation of thiol groups. Protective and protein purification agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5302