Learn More
Medchemexpress LLC Dithizone 5g
SDP
Supplier:  Medchemexpress LLC HYW1107935G
Encompass_Preferred
Dithizone can be toxic to pancreatic islet cells and cause experimental diabetes Dithizone can be used for in vitro in vivo staining of transplanted islets and cytoplasmic granules of bone marrow cells Dithizone can be used as a chelating agent and colorimetric indicator for the detection and extraction of various heavy metals including lead and mercury in analytical and biochemical applications Dithizone forms stable colored complexes with metal ions and can be quantitatively analyzed by methods such as spectrophotometry[1][2]
Catalog No. 50-002-06639