Learn More
Medchemexpress LLC Dithranol | 1143-38-0 | ≥98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB073810MM/1ML
Encompass_Preferred
Dithranol, also known as Anthralin, is an anthraquinone derivative with potent anti-psoriatic effects. It functions by inhibiting DNA replication and repair mechanisms within cells. This compound is intended for research use only.
  • Inhibits DNA replication and repair
  • Potent anti-psoriatic effects
  • Inhibits replication in T98G cells with a GI50 of 1.1 μM
  • Increases mRNA production of interleukin-10 receptors in HaCaT cells
Catalog No. 50-004-06757