Learn More
Medchemexpress LLC Ditiocarb sodium | 148-18-5 | 99.7% | 50 G
SDP
Supplier:  Medchemexpress LLC HYB163750G
Encompass_Preferred
Ditiocarb sodium (Sodium diethyldithiocarbamate) is an orally active copper reagent. It exhibits activities such as antioxidation, chelation, anti-tumor effects, immunomodulation, and anti-HIV properties. It can be used in the research of tumors, inflammatory, and immune-related diseases.
  • Orally active copper reagent.
  • Exhibits antioxidation, chelation, anti-tumor effects, immunomodulation, and anti-HIV properties.
  • Used in research of tumors, inflammatory, and immune-related diseases.
  • Can act as an accelerator in the cementation of copper from copper sulfate solution on zinc.
  • Reacts with Cu2+ solution to form a copper diethyldithiocarbamate complex, increasing the rate of copper cementation.
  • Inhibits Cisplatin-induced nephrotoxicity and gastrointestinal toxicity in rats.
  • Exhibits antitumor activity in a mouse skin tumor model.
Catalog No. 50-004-13294