Learn More
Medchemexpress LLC Ditiocarb sodium (Sodium diethyldithiocarbamate) | 148-18-5 | 99.2% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB1637A10G
Encompass_Preferred
Ditiocarb sodium is an orally active copper reagent that exhibits activities such as antioxidation, chelation, anti-tumor effects, immunomodulation, and anti-HIV properties. It can be used in the research of tumors, inflammatory, and immune-related diseases.
  • Suitable for high-speed applications
  • 50 mL capacity
  • Leak-resistant cap
  • Inhibits Cisplatin-induced nephrotoxicity and gastrointestinal toxicity
  • Exhibits antitumor activity in a mouse skin tumor model
Catalog No. 50-004-13292