Learn More
Medchemexpress LLC Ditiocarb sodium (Sodium diethyldithiocarbamate) | 148-18-5 | MFCD00066667 | 99.2% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB1637A5G
Encompass_Preferred
Ditiocarb sodium is an orally active copper reagent that exhibits various activities including antioxidation, chelation, anti-tumor effects, immunomodulation, and anti-HIV properties. It is suitable for research in tumors, inflammatory, and immune-related diseases.
  • Acts as an accelerator in the cementation of copper from copper sulfate solution on zinc by forming a copper diethyldithiocarbamate complex.
  • Inhibits Cisplatin-induced nephrotoxicity and gastrointestinal toxicity in rats.
  • Exhibits antitumor activity in a mouse skin tumor model.
Catalog No. 50-004-13276